1,4-Diethyl-2-bromobenzene-1,4-dicarboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR303074-10G
OR303074-1G
OR303074-5G
APOSOR303074-10GEA
894
EUR
APOSOR303074-10G
APOSOR303074-1G
APOSOR303074-5G
1,4-Diethyl-2-bromobenzene-1,4-dicarboxylate
1,4-Diethyl-2-bromobenzene-1,4-dicarboxylate
Formel:
C₁₂H₁₃BrO₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD12913608 CAS-Nummer: 154239-21-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...