Diethyl-2-{[(6-methoxypyridin-3-yl)amino]methylene}malonate

Lieferant: Apollo Scientific

Synonyme: DIethyl 2-{[(6-methoxypyridin-3-yl)amino]methylene}malonate

OR303031-1G OR303031-500MG OR303031-5G
APOSOR303031-1GEA 180 EUR
APOSOR303031-1G APOSOR303031-500MG APOSOR303031-5G
Diethyl-2-{[(6-methoxypyridin-3-yl)amino]methylene}malonate
Diethyl-2-{[(6-methoxypyridin-3-yl)amino]methylene}malonate
Formel: C₁₄H₁₈N₂O₅
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08457989
CAS-Nummer: 53241-90-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR