2-Hydrazino-4-methoxypyrimidine

Lieferant: Apollo Scientific

OR302395-1G OR302395-5G
APOSOR302395-1GEA 300 EUR
APOSOR302395-1G APOSOR302395-5G
2-Hydrazino-4-methoxypyrimidine
2-Hydrazino-4-methoxypyrimidine
Formel: C₅H₈N₄O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02734394
CAS-Nummer: 89181-80-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR