4-Methyl-5-nitro-1H-pyrazole
Lieferant: Apollo Scientific
|
Achtung
|
OR302333-1G
OR302333-250MG
OR302333-5G
APOSOR302333-1GEA
65.4
EUR
APOSOR302333-1G
APOSOR302333-250MG
APOSOR302333-5G
4-Methyl-5-nitro-1H-pyrazole
4-Methyl-5-nitro-1H-pyrazole
Formel:
C₄H₅N₃O₂ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD08685901 CAS-Nummer: 38858-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...