Methyl-5-bromo-2-methylbenzoate

Lieferant: Apollo Scientific
Achtung

OR301032-100G OR301032-25G
APOSOR301032-100GEA 112 EUR
APOSOR301032-100G APOSOR301032-25G
Methyl-5-bromo-2-methylbenzoate
Methyl-5-bromo-2-methylbenzoate
Formel: C₉H₉BrO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09839555
CAS-Nummer: 79669-50-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR