3-Methoxy-o-toluidin

Lieferant: Apollo Scientific

OR301008-100G OR301008-1G
APOSOR301008-100GEA 286 EUR
APOSOR301008-100G APOSOR301008-1G
3-Methoxy-o-toluidin
3-Methoxy-o-toluidin
Formel: C₈H₁₁NO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06412568
CAS-Nummer: 19500-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR