2-Chloroisonicotinic acid hydrazide
Lieferant: Apollo Scientific
|
Achtung
|
OR29934-1G
OR29934-250MG
OR29934-5G
APOSOR29934-1GEA
0
EUR
APOSOR29934-1G
APOSOR29934-250MG
APOSOR29934-5G
2-Chloroisonicotinic acid hydrazide
2-Chloroisonicotinic acid hydrazide
Formel:
C₆H₆ClN₃O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00221428 CAS-Nummer: 58481-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...