5-Bromo-4,6-dichloropyrimidin-2-amine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-5-bromo-4,6-dichloropyrimidine
OR2987-1G
OR2987-250MG
APOSOR2987-1GEA
119
EUR
APOSOR2987-1G
APOSOR2987-250MG
5-Bromo-4,6-dichloropyrimidin-2-amine
5-Bromo-4,6-dichloropyrimidin-2-amine
Formel:
C₄H₂BrCl₂N₃ Schmelzpunkt: 232…236 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00275455 CAS-Nummer: 7781-26-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...