1-(5-Methyl-3-phenyl-4-isoxazolyl)-1-ethanone 97%
Lieferant: Apollo Scientific
Synonyme:
4-Acetyl-5-methyl-3-phenylisoxazole
OR29752-1G
OR29752-5G
APOSOR29752-1GEA
0
EUR
APOSOR29752-1G
APOSOR29752-5G
1-(5-Methyl-3-phenyl-4-isoxazolyl)-1-ethanone 97%
1-(5-Methyl-3-phenyl-4-isoxazolyl)-1-ethanone
Formel:
C₁₂H₁₁NO₂ Molekulargewicht: 201.23 g/mol Schmelzpunkt: 56…58 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00173901 CAS-Nummer: 19212-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...