Methyl 5-bromo-2-hydroxy-3-nitrobenzenecarboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 5-bromo-2-hydroxy-3-nitrobenzoate
OR2935-1G
OR2935-5G
APOSOR2935-1GEA
65.4
EUR
APOSOR2935-1G
APOSOR2935-5G
Methyl 5-bromo-2-hydroxy-3-nitrobenzenecarboxylate
Methyl-5-bromo-2-hydroxy-3-nitrobenzenecarboxylate
Formel:
C₈H₆BrNO₅ Schmelzpunkt: 146…148 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00458681 CAS-Nummer: 91983-31-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...