5-Bromnicotinsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromo-5-pyridine carboxylic acid, 5-Bromopyridine-3-carboxylic acid, 5-Bromo-3-pyridinecarboxylic acid
OR28916-100G
OR28916-1KG
OR28916-500G
APOSOR28916-100GEA
55.1
EUR
APOSOR28916-100G
APOSOR28916-1KG
APOSOR28916-500G
5-Bromnicotinsäure 98%
5-Bromnicotinsäure
Formel:
C₆H₄BrNO₂ Schmelzpunkt: 178…186 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00009783 CAS-Nummer: 20826-04-4 EINECS: 244-065-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...