Histamin Dihydrochlorid

Lieferant: Apollo Scientific
Gefahr

Synonyme: Histamine 2HCl, 4-(2-Aminoethyl)-1H-imidazole dihydrochloride, 2-(1H-Imidazol-4-yl)ethan-1-amine dihydrochloride

OR28842-25G OR28842-5G OR28842-100G
APOSOR28842-25GEA 65.4 EUR
APOSOR28842-25G APOSOR28842-5G APOSOR28842-100G
Histamin Dihydrochlorid
Histamin Dihydrochlorid
Formel: C₅H₁₁Cl₂N₃
Schmelzpunkt: 249…252 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00012703
CAS-Nummer: 56-92-8
EINECS: 200-298-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR