Methyl mercaptan sodium salt

Lieferant: Apollo Scientific
Gefahr

Synonyme: Sodium methanethiolate, Sodium thiomethoxide, Sodium methylsulphanide, Methanethiol sodium salt

OR28823-25G OR28823-5G
APOSOR28823-25GEA 236 EUR
APOSOR28823-25G APOSOR28823-5G
Methyl mercaptan sodium salt
Methyl mercaptan sodium salt
Formel: CH₃NaS
Molekulargewicht: 70.08 g/mol
Flammpunkt: 27 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00174316
CAS-Nummer: 5188-07-8
EINECS: 225-969-9
UN-Nummer: 2921
ADR: 8+4.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR