6-Amino-7-deazapurine hydrogen sulphate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Amino-7H-pyrrolo[2,3-d]pyrimidine sulphate
OR2834-1G
OR2834-5G
OR2834-25G
APOSOR2834-1GEA
163
EUR
APOSOR2834-1G
APOSOR2834-5G
APOSOR2834-25G
6-Amino-7-deazapurine hydrogen sulphate
6-Amino-7-deazapurine hydrogen sulphate
Formel:
C₆H₈N₄O₄S Molekulargewicht: 232.214 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09787504 CAS-Nummer: 769951-32-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...