Methyl-1H-benzimidazol-5-carboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzimidazole-5-carboxylic acid methyll ester

OR25948-1G OR25948-5G
APOSOR25948-1GEA 21.9 EUR
APOSOR25948-1G APOSOR25948-5G
Methyl-1H-benzimidazol-5-carboxylat
Methyl benzimidazole-5-carboxylate
Formel: C₉H₈N₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03407310
CAS-Nummer: 26663-77-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR