1-Benzyl-1H-indole-3-carbaldehyde 95%

Lieferant: Apollo Scientific
Achtung

OR22348-1G OR22348-250MG
APOSOR22348-1GEA 306 EUR
APOSOR22348-1G APOSOR22348-250MG
1-Benzyl-1H-indole-3-carbaldehyde 95%
1-Benzyl-1H-indole-3-carbaldehyde
Formel: C₁₆H₁₃NO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00022894
CAS-Nummer: 10511-51-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR