2-Brom-4-methyl-6-nitroanilin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-6-nitro-p-toluidine, 4-Amino-3-bromo-5-nitrotoluene
OR22152-25G
OR22152-5G
APOSOR22152-25GEA
53.8
EUR
APOSOR22152-25G
APOSOR22152-5G
2-Brom-4-methyl-6-nitroanilin
2-Brom-4-methyl-6-nitroanilin
Formel:
C₇H₇BrN₂O₂ Schmelzpunkt: 64…66 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00209452 CAS-Nummer: 827-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...