3-(2,4-Dimethylphenylcarbamoyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR2097-1G
OR2097-5G
OR2097-25G
APOSOR2097-1GEA
50.9
EUR
APOSOR2097-1G
APOSOR2097-5G
APOSOR2097-25G
3-(2,4-Dimethylphenylcarbamoyl)phenylboronsäure 98%
3-(2,4-Dimethylphenylcarbamoyl)phenylboronsäure
Formel:
C₁₅H₁₆BNO₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09027261 CAS-Nummer: 957060-98-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...