3-(N,N-Dimethylamino)phenylboronsäure 96%
Lieferant: Apollo Scientific
Synonyme:
3-Dimethylaminophenylboronic acid, 3-(Dimethylamino)benzeneboronic acid
OR2015-25G
OR2015-5G
APOSOR2015-25GEA
260
EUR
APOSOR2015-25G
APOSOR2015-5G
3-(N,N-Dimethylamino)phenylboronsäure 96%
3-(N,N-Dimethylamino)phenylboronsäure
Formel:
C₈H₁₂BNO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01318996 CAS-Nummer: 178752-79-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...