L(+)-α-Phenylglycin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
H-Phg-OH, L-(+)-2-Phenylglycine
OR19581-100G
OR19581-25G
OR19581-500G
APOSOR19581-100GEA
24.8
EUR
APOSOR19581-100G
APOSOR19581-25G
APOSOR19581-500G
L(+)-α-Phenylglycin 97%
L(+)-α-Phenylglycin
Formel:
C₈H₉NO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00064403 CAS-Nummer: 2935-35-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...