N-Benzoyl-N-phenylhydroxylamine 98%

Lieferant: Apollo Scientific
Achtung

OR18574-25G OR18574-5G OR18574-1G
APOSOR18574-25GEA 141 EUR
APOSOR18574-25G APOSOR18574-5G APOSOR18574-1G
N-Benzoyl-N-phenylhydroxylamine 98%
N-Benzoyl-N-phenylhydroxylamine
Formel: C₁₃H₁₁NO₂
Schmelzpunkt: 118…123 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00002111
CAS-Nummer: 304-88-1
EINECS: 206-158-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR