5-Bromo-2-methylpyridin-3-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-5-bromo-2-methylpyridine

OR18558-5G OR18558-10G
APOSOR18558-5GEA 60.9 EUR
APOSOR18558-5G APOSOR18558-10G
5-Bromo-2-methylpyridin-3-amine
5-Bromo-2-methylpyridin-3-amine
Formel: C₆H₇BrN₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09031418
CAS-Nummer: 914358-73-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR