2,5-Dibromphenol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,3-Dibromo-4-hydroxybenzene
OR18409-1G
OR18409-5G
OR18409-10G
APOSOR18409-1GEA
21.9
EUR
APOSOR18409-1G
APOSOR18409-5G
APOSOR18409-10G
2,5-Dibromphenol 98%
2,5-Dibromphenol
Formel:
C₆H₄Br₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01851373 CAS-Nummer: 28165-52-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...