(Quinolin-2-ylmethyl)amine hydrochloride 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Aminomethyl)quinoline hydrochloride, Quinolin-2-ylmethanamine hydrochloride
OR18031-250MG
OR18031-100MG
OR18031-1G
APOSOR18031-250MGEA
313
EUR
APOSOR18031-250MG
APOSOR18031-100MG
APOSOR18031-1G
(Quinolin-2-ylmethyl)amine hydrochloride 97%
(Quinolin-2-ylmethyl)amine hydrochloride
Formel:
C₁₀H₁₁ClN₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09966142 CAS-Nummer: 861036-67-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...