Ethyl-4-amino-2-chlorbenzoat 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR17937-1G
OR17937-500MG
APOSOR17937-1GEA
24.8
EUR
APOSOR17937-1G
APOSOR17937-500MG
Ethyl-4-amino-2-chlorbenzoat 95+%
Ethyl-4-amino-2-chlorbenzoat
Formel:
C₉H₁₀ClNO₂ Schmelzpunkt: 109…111 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD09034640 CAS-Nummer: 16017-69-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...