4-Bromo-1,1,3,3,6-pentamethyl-7-nitro-2,3-dihydro-1H-inden-5-ylamine

Lieferant: Apollo Scientific

Synonyme: 5-Amino-4-bromo-7-nitro-1,1,3,3,6-pentamethylindane

OR16156-1G OR16156-5G
APOSOR16156-1GEA 105 EUR
APOSOR16156-1G APOSOR16156-5G
4-Bromo-1,1,3,3,6-pentamethyl-7-nitro-2,3-dihydro-1H-inden-5-ylamine
4-Bromo-1,1,3,3,6-pentamethyl-7-nitro-2,3-dihydro-1H-inden-5-ylamine
Formel: C₁₄H₁₉BrN₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11100123
CAS-Nummer: 1210378-97-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR