N-Methyl-1-(methylsulphonyl)piperidin-4-amine
Lieferant: Apollo Scientific
Synonyme:
4-(Methylamino)-1-(methylsulphonyl)piperidine
OR16035-1G
OR16035-50MG
OR16035-100MG
OR16035-250MG
OR16035-500MG
APOSOR16035-1GEA
357
EUR
APOSOR16035-1G
APOSOR16035-50MG
APOSOR16035-100MG
APOSOR16035-250MG
APOSOR16035-500MG
N-Methyl-1-(methylsulphonyl)piperidin-4-amine
N-Methyl-1-(methylsulphonyl)piperidin-4-amine
Formel:
C₇H₁₆N₂O₂S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08061933 CAS-Nummer: 438585-61-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...