2-Chloro-5-nitro-1H-1,3-benzimidazole 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-6-nitro-1H-benzo[d]imidazole
OR15280-10G
OR15280-1G
OR15280-5G
OR15280-25G
APOSOR15280-10GEA
894
EUR
APOSOR15280-10G
APOSOR15280-1G
APOSOR15280-5G
APOSOR15280-25G
2-Chloro-5-nitro-1H-1,3-benzimidazole 95%
2-Chloro-5-nitro-1H-1,3-benzimidazole
Formel:
C₇H₄ClN₃O₂ Schmelzpunkt: 225…227 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00769824 CAS-Nummer: 5955-72-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...