6-[(tert-Butoxycarbonyl)amino]nicotinic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Aminonicotinic acid, 6-BOC protected, 6-((tert-Butoxycarbonyl)amino)nicotinic acid

OR15274-1G OR15274-250MG
APOSOR15274-1GEA 130 EUR
APOSOR15274-1G APOSOR15274-250MG
6-[(tert-Butoxycarbonyl)amino]nicotinic acid
6-[(tert-Butoxycarbonyl)amino]nicotinic acid
Formel: C₁₁H₁₄N₂O₄
Molekulargewicht: 238.25 g/mol
Schmelzpunkt: 292…296 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02682388
CAS-Nummer: 231958-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR