1-(4-Methoxyphenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dimethyl-1-(4-methoxyphenyl)-1H-pyrazole-4-carboxylic acid

OR15209-10G OR15209-1G OR15209-5G
APOSOR15209-10GEA 1350 EUR
APOSOR15209-10G APOSOR15209-1G APOSOR15209-5G
1-(4-Methoxyphenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid
1-(4-Methoxyphenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid
Formel: C₁₃H₁₄N₂O₃
Schmelzpunkt: 180…183 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08310579
CAS-Nummer: 1001779-87-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR