H-Phe(4-tBu)-OH 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: H-p-tBu-Phe-OH

OR14740-1G OR14740-5G
APOSOR14740-1GEA 58 EUR
APOSOR14740-1G APOSOR14740-5G
H-Phe(4-tBu)-OH 96%
H-Phe(4-tBu)-OH
Formel: C₁₃H₁₉NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02259528
CAS-Nummer: 82372-74-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR