4-Methyl-D-phenylalanine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
H-D-Phe(4-Me)-OH, H-p-Me-D-Phe-OH
OR14704-5G
OR14704-1G
APOSOR14704-5GEA
63.8
EUR
APOSOR14704-5G
APOSOR14704-1G
4-Methyl-D-phenylalanine 98%
4-Methyl-D-phenylalanine
Formel:
C₁₀H₁₃NO₂ Schmelzpunkt: 84…88 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00152137 CAS-Nummer: 49759-61-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...