(2R)-2-[(tert-Butoxycarbonyl)amino]-3-(1,3-thiazol-4-yl)propanoic acid

Lieferant: Apollo Scientific

OR14635-1G OR14635-250MG
APOSOR14635-1GEA 176 EUR
APOSOR14635-1G APOSOR14635-250MG
(2R)-2-[(tert-Butoxycarbonyl)amino]-3-(1,3-thiazol-4-yl)propanoic acid
(2R)-2-[(tert-Butoxycarbonyl)amino]-3-(1,3-thiazol-4-yl)propanoic acid
Formel: C₁₁H₁₆N₂O₄S
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00079680
CAS-Nummer: 134107-69-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR