2-Bromo-N-fmoc-L-phenylalanine
Lieferant: Apollo Scientific
Synonyme:
Fmoc-Phe(2-Br)-OH
OR14554-1G
OR14554-5G
APOSOR14554-1GEA
33.5
EUR
APOSOR14554-1G
APOSOR14554-5G
2-Bromo-N-fmoc-L-phenylalanine
2-Bromo-N-fmoc-L-phenylalanine
Formel:
C₂₄H₂₀BrNO₄ Molekulargewicht: 466.331 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01311743 CAS-Nummer: 220497-47-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...