5-(2-Aminoethyl)-1H-1,2,4-triazol-3-amine dihydrochloride

Lieferant: Apollo Scientific
Achtung

OR13428-100MG OR13428-250MG
APOSOR13428-100MGEA 116 EUR
APOSOR13428-100MG APOSOR13428-250MG
5-(2-Aminoethyl)-1H-1,2,4-triazol-3-amine dihydrochloride
5-(2-Aminoethyl)-1H-1,2,4-triazol-3-amine dihydrochloride
Formel: C₄H₁₁Cl₂N₅
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD20441445
CAS-Nummer: 99839-36-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR