3-Amino-4-chlorobenzeneboronic acid hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR13163-1G OR13163-5G OR13163-10G
APOSOR13163-1GEA 75.4 EUR
APOSOR13163-1G APOSOR13163-5G APOSOR13163-10G
3-Amino-4-chlorobenzeneboronic acid hydrochloride 95%
3-Amino-4-chlorobenzeneboronic acid hydrochloride
Formel: C₆H₇BClNO₂·HCl
Schmelzpunkt: 278…288 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04115643
CAS-Nummer: 850568-45-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR