6-Chlor-2-tetralon
Lieferant: Apollo Scientific
OR12932-1G
OR12932-5G
APOSOR12932-1GEA
97.3
EUR
APOSOR12932-1G
APOSOR12932-5G
6-Chlor-2-tetralon
Formel:
C₁₀H₉ClO Schmelzpunkt: 60…65 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03788443 CAS-Nummer: 17556-18-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...