3,5-Dimethyl-4-isoxazolcarbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Dimethylisoxasole-4-carboxylic acid
OR1283-10G
OR1283-25G
OR1283-5G
APOSOR1283-10GEA
77
EUR
APOSOR1283-10G
APOSOR1283-25G
APOSOR1283-5G
3,5-Dimethyl-4-isoxazolcarbonsäure
3,5-Dimethylisoxazole-4-carboxylic acid
Formel:
C₆H₇NO₃ Schmelzpunkt: 140…142 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00051657 CAS-Nummer: 2510-36-3 EINECS: 219-724-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...