4-Phenoxypiperidin Hydrochlorid

Lieferant: Apollo Scientific
Achtung

OR12597-1G OR12597-250MG
APOSOR12597-1GEA 29 EUR
APOSOR12597-1G APOSOR12597-250MG
4-Phenoxypiperidin Hydrochlorid
4-Phenoxypiperidin Hydrochlorid
Formel: C₁₁H₁₆ClNO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02684106
CAS-Nummer: 3413-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR