5-(4-Chlorphenyl)oxazol

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(4-Chlorophenyl)-1,3-oxazole

OR12571-1G OR12571-5G
APOSOR12571-1GEA 104 EUR
APOSOR12571-1G APOSOR12571-5G
5-(4-Chlorphenyl)oxazol
5-(4-Chlorophenyl)oxazole
Formel: C₉H₆ClNO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09258870
CAS-Nummer: 1008-94-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR