2-Brom-2,6-dichlortoluol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-3,5-dichloro-2-methylbenzene
OR11949-1G
OR11949-250MG
APOSOR11949-1GEA
174
EUR
APOSOR11949-1G
APOSOR11949-250MG
2-Brom-2,6-dichlortoluol
2-Brom-2,6-dichlortoluol
Formel:
C₇H₅BrCl₂ Molekulargewicht: 239.93 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09801039 CAS-Nummer: 115615-19-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...