trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2E)-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, (E)-Ferulic acid, Trans-3-(4-hydroxy-3-methoxyphenyl)acrylic acid, trans-Ferulic acid

OR1171-100G OR1171-25G OR1171-500G
APOSOR1171-100GEA 27.7 EUR
APOSOR1171-100G APOSOR1171-25G APOSOR1171-500G
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) 98%
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid
Formel: C₁₀H₁₀O₄
Schmelzpunkt: 169…173 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00004400
CAS-Nummer: 537-98-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR