3,5-Dibromanilin Hydrochlorid

Lieferant: Apollo Scientific
Achtung

OR11689-1G OR11689-5G
APOSOR11689-1GEA 29 EUR
APOSOR11689-1G APOSOR11689-5G
3,5-Dibromanilin Hydrochlorid
3,5-Dibromanilin Hydrochlorid
Formel: C₆H₆Br₂ClN
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09800855
CAS-Nummer: 188349-40-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR