4-Hydroxycumarin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Hydroxy-2-benzopyrone
OR1147-2.5KG
OR1147-500G
OR1147-100G
APOSOR1147-2.5KGEA
214
EUR
APOSOR1147-2.5KG
APOSOR1147-500G
APOSOR1147-100G
4-Hydroxycumarin 98%
4-Hydroxycumarin
Formel:
C₉H₆O₃ Schmelzpunkt: 216 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00006856 CAS-Nummer: 1076-38-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...