tert-Butyl-4-brom-2-nitrobenzoat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 4-bromo-2-nitrobenzoate, 4-Bromobenzoic acid tert-butyl ester
OR11452-1G
OR11452-5G
APOSOR11452-1GEA
21.9
EUR
APOSOR11452-1G
APOSOR11452-5G
tert-Butyl-4-brom-2-nitrobenzoat 98%
tert-Butyl-4-brom-2-nitrobenzoat
Formel:
C₁₁H₁₂BrNO₄ Molekulargewicht: 302.12 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09475812 CAS-Nummer: 890315-72-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...