2-Acetyl-1-naphthol

Lieferant: Apollo Scientific

OR1123-100G OR1123-25G
APOSOR1123-100GEA 294 EUR
APOSOR1123-100G APOSOR1123-25G
2-Acetyl-1-naphthol
2-Acetyl-1-naphthol
Formel: C₁₂H₁₀O₂
Schmelzpunkt: 98…100 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00003963
CAS-Nummer: 711-79-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR