5-Amino-1-tert-butyl-1H-pyrazole-4-carbonitrile 95%

Lieferant: Apollo Scientific

Synonyme: 5-Amino-1-tert-butyl-1h-pyrazole-4-carbonitrile, 5-Amino-1-(tert-butyl)-1H-pyrazole-4-carbonitrile

OR111100-1G OR111100-5G
APOSOR111100-1GEA 101 EUR
APOSOR111100-1G APOSOR111100-5G
5-Amino-1-tert-butyl-1H-pyrazole-4-carbonitrile 95%
5-Amino-1-tert-butyl-1H-pyrazole-4-carbonitrile
Formel: C₈H₁₂N₄
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00128289
CAS-Nummer: 158001-28-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR