1H-Indole-2-carbonyl chloride

Lieferant: Apollo Scientific

OR111006-1G OR111006-5G
APOSOR111006-1GEA 241 EUR
APOSOR111006-1G APOSOR111006-5G
1H-Indole-2-carbonyl chloride
1H-Indole-2-carbonyl chloride
Formel: C₉H₆ClNO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09833544
CAS-Nummer: 58881-45-1
UN-Nummer: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR