8-Methylimidazo[1,2-a]pyridin

Lieferant: Apollo Scientific

OR110909-1G OR110909-5G
APOSOR110909-1GEA 147 EUR
APOSOR110909-1G APOSOR110909-5G
8-Methylimidazo[1,2-a]pyridin
8-Methylimidazo[1,2-a]pyridin
Formel: C₈H₈N₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD05663805
CAS-Nummer: 874-10-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR