2-(2-Chlorophenyl)glycine

Lieferant: Apollo Scientific
Achtung

Synonyme: H-DL-Phg(2-Cl)-OH, 2-Amino-2-(2-chlorophenyl)acetic Acid, (±)-2-Chlorophenylglycine

OR11056-100G OR11056-25G
APOSOR11056-100GEA 24.8 EUR
APOSOR11056-100G APOSOR11056-25G
2-(2-Chlorophenyl)glycine
2-(2-Chlorophenyl)glycine
Formel: C₈H₈ClNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00049324
CAS-Nummer: 88744-36-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR